Glycolaldehyde diethyl acetal

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Hydroxyacetaldehyde diethyl acetal, 2,2-Diethoxyethanol

OR16597-100G OR16597-25G OR16597-5G
APOSOR16597-100GEA 2240 DKK
APOSOR16597-100G APOSOR16597-25G APOSOR16597-5G
Glycolaldehyde diethyl acetal
Glycolaldehyde diethyl acetal
Formel: HOCH₂CH(OCH₂CH₃)₂
Kogepunkt: 166…167 °C
Flammepunkt: 62 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00051486
CAS-nummer: 621-63-6
EINECS: 210-697-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR