Fmoc-β-(2-thienyl)-D-alanine 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-N-Fmoc-2-thienylalanine, Fmoc-D-Thi-OH, Fmoc-D-3-(2-Thienyl)-Ala-OH, Fmoc-3-D-Ala(2-thienyl)-OH, Fmoc-β-(2-thienyl)-D-Ala-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-2-yl)propanoic acid, Fmoc-D-2-thienylalanine

OR14645-5G
APOSOR14645-5GEA 676 DKK
APOSOR14645-5G
Fmoc-β-(2-thienyl)-D-alanine 96%
Fmoc-β-(2-thienyl)-D-alanine
Formel: C₂₂H₁₉NO₄S
Smeltepunkt: 168…171 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00065677
CAS-nummer: 201532-42-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR