4-(3-Nitrobenzyl)piperazine, N1-BOC protected 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: tert-Butyl 4-(3-nitrobenzyl)piperazine-1-carboxylate

OR1461-1G
APOSOR1461-1GEA 709 DKK
APOSOR1461-1G
4-(3-Nitrobenzyl)piperazine, N1-BOC protected 97%
4-(3-Nitrobenzyl)piperazine, N1-BOC protected
Formel: C₁₆H₂₃N₃O₄
Molekylvægt: 321.372 g/mol
Smeltepunkt: 84…85 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06797469
CAS-nummer: 203047-33-0

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR