N-Hydroxy-4-methoxybenzenecarboximidoyl chloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: alpha-Chloro-4-methoxybenzaldoxime

OR14213-1G
APOSOR14213-1GEA 818 DKK
APOSOR14213-1G
N-Hydroxy-4-methoxybenzenecarboximidoyl chloride
N-Hydroxy-4-methoxybenzenecarboximidoyl chloride
Formel: C₈H₈ClNO₂
Smeltepunkt: 84…89 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01924588
CAS-nummer: 38435-51-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR