5-(3-Nitrophenyl)oxazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR1252-5G
APOSOR1252-5GEA 1090 DKK
APOSOR1252-5G
5-(3-Nitrophenyl)oxazole
5-(3-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00085147
CAS-nummer: 89808-77-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR