1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR111300-5G OR111300-1G
APOSOR111300-5GEA 1490 DKK
APOSOR111300-5G APOSOR111300-1G
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
Formel: C₁₉H₂₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00428365
CAS-nummer: 59662-38-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR