1-Methylimidazole-4-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Methyl-1H-imidazole-4-carboxylic acid

OR10542-1G OR10542-5G OR10542-25G
APOSOR10542-1GEA 164 DKK
APOSOR10542-1G APOSOR10542-5G APOSOR10542-25G
1-Methylimidazole-4-carboxylic acid
1-Methylimidazole-4-carboxylic acid
Formel: C₅H₆N₂O₂
MDL Number: MFCD02179560
CAS-nummer: 41716-18-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR