Indol-7-carbaldehyd

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1H-Indole-7-carbaldehyde, Indole-7-carboxaldehyde

OR10490-250MG
APOSOR10490-250MGEA 164 DKK
APOSOR10490-250MG
Indol-7-carbaldehyd
Indol-7-carbaldehyd
Formel: C₉H₇NO
Smeltepunkt: 87…91 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01318152
CAS-nummer: 1074-88-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR