5-Formyl-2-thiophenboronsyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

OR10486-100G OR10486-25G OR10486-5G
APOSOR10486-100GEA 1620 DKK
APOSOR10486-100G APOSOR10486-25G APOSOR10486-5G
5-Formyl-2-thiophenboronsyre
5-Formyl-2-thiophenboronsyre
Formel: C₅H₅BO₃S
Smeltepunkt: 132…135 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02093666
CAS-nummer: 4347-33-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR