D(+)-Prolin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

OR0718-100G OR0718-25G OR0718-5G
APOSOR0718-100GEA 1160 DKK
APOSOR0718-100G APOSOR0718-25G APOSOR0718-5G
D(+)-Prolin
D(+)-Prolin

Chiral intermediate for pharmaceuticals and agrochemicals.

Formel: C₅H₉NO₂
Smeltepunkt: 223 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064317
CAS-nummer: 344-25-2
EINECS: 206-452-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR