3-(Cyclopentyloxy)-4-methoxybenzaldehyde

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(Cyclopentyloxy)-4-formylanisole

OR0323-50MG OR0323-100MG
APOSOR0323-50MGEA 164 DKK
APOSOR0323-50MG APOSOR0323-100MG
3-(Cyclopentyloxy)-4-methoxybenzaldehyde
3-(Cyclopentyloxy)-4-methoxybenzaldehyde
Formel: C₁₃H₁₆O₃
Kogepunkt: 124…126 °C (0,05 mm Hg)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00673136
CAS-nummer: 67387-76-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR