Sodium thiophosphate hydrate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

IN3322-25G IN3322-1G IN3322-5G
APOSIN3322-25GEA 1910 DKK
APOSIN3322-25G APOSIN3322-1G APOSIN3322-5G
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate
Formel: H₂Na₃O₄PS
Smeltepunkt: 56…58 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00041513
CAS-nummer: 10489-48-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR