Jern (III) chlorid hexahydrat ACS

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Iron trichloride hexahydrate, Ferric chloride hexahydrate

IN2065-1KG IN2065-5KG
APOSIN2065-1KGEA 1660 DKK
APOSIN2065-1KG APOSIN2065-5KG
Jern (III) chlorid hexahydrat ACS
Jern (III) chlorid hexahydrat
Formel: FeCl₃·6H₂O
Kogepunkt: 280…285 °C
Smeltepunkt: 37 °C
Massefylde: 1,82 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149712
CAS-nummer: 10025-77-1
UN nummer: 3260
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR