Bismuthtriacetat 99,8%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate

IN1237-25G
APOSIN1237-25GEA 545 DKK
APOSIN1237-25G
Bismuthtriacetat 99,8%
Bismuthtriacetat
Formel: (H₃CCOO)₃Bi
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015629
CAS-nummer: 22306-37-2
EINECS: 244-904-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR