Pepsin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIP1008-100G BIP1008-1KG BIP1008-500G
APOSBIP1008-100GEA 985 DKK
APOSBIP1008-100G APOSBIP1008-1KG APOSBIP1008-500G
Pepsin
Enzymer

Bestil nu

Specifikationer

Enzym navn Pepsin
Environmentally Preferable
Kilde
Species
MDL No. MFCD00081840
Storage conditions Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR