β-Thionicotinamide adenine dinucleotide, reduced form disodium salt 92+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Thio-NADH

BIB5005-100MG BIB5005-250MG BIB5005-50MG
APOSBIB5005-100MGEA 915 DKK
APOSBIB5005-100MG APOSBIB5005-250MG APOSBIB5005-50MG
β-Thionicotinamide adenine dinucleotide, reduced form disodium salt 92+%
β-Thionicotinamide adenine dinucleotide, reduced form disodium salt

Purity determined enzymatic method (ADH)

Formel: C₂₁H₂₇N₇O₁₃P₂S·2Na
Opbevaringstemperatur: Fryser
MDL Number: MFCD00133854
CAS-nummer: 1921-48-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR