D(+)-Biotin 99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

BIB0603-1G BIB0603-5G BIB0603-10G
APOSBIB0603-1GEA 218 DKK
APOSBIB0603-1G APOSBIB0603-5G APOSBIB0603-10G
D(+)-Biotin 99%
D(+)-Biotin
Formel: C₁₀H₁₆N₂O₃S
Smeltepunkt: 231…233 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005541
CAS-nummer: 58-85-5
EINECS: 200-399-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR