Calciumchlorid dihydrat

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Calcium Chloride Dihydrate, Calcium dichloride

BIA4689-250G
APOSBIA4689-250GEA 0 DKK
APOSBIA4689-250G
Calciumchlorid dihydrat
Calciumchlorid dihydrat
Formel: CaCl₂·2H₂O
Molekylvægt: 147.01 g/mol
Kogepunkt: 1600
Smeltepunkt: 175 °C
Massefylde: 1.85 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149613
CAS-nummer: 10035-04-8
Merck Index: 13,01660

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR