D(+)-Prolin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

BIA0718-100G BIA0718-25G
APOSBIA0718-100GEA 382 DKK
APOSBIA0718-100G APOSBIA0718-25G
D(+)-Prolin
D(+)-Prolin
Formel: C₅H₉NO₂
Smeltepunkt: 223 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064317
CAS-nummer: 344-25-2
EINECS: 206-452-7

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR