Chemical resistant shoe covers, MICROGARD® 2500, model 400
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WH25B-00400-00
ANSEWH25B-00400-00EA 7810 DKK
ANSEWH25B-00400-00
Chemical resistant shoe covers, MICROGARD® 2500, model 400
Engangsbeklædning
These over shoes are made of microporous PP film with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
  • Elasticated opening
  • Bound seams
  • Fit shoe size 42 - 46