Chemical resistant boot covers, AlphaTec® 2000, Model 406
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WH20B-00406-00
ANSEWH20B-00406-00EA 3660 DKK
ANSEWH20B-00406-00
Chemical resistant boot covers, AlphaTec® 2000, Model 406
Engangsbeklædning
Superior breathable microporous laminate technology.
  • Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
  • Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
  • Optimised fit, improving wearer comfort and safety
  • Low Linting
Boot covers feature tie fastening, elastic to top of boot, and bound seams. Covers are manufactured from non-woven microporous polyethylene laminate material.
Recommended for cleanroom preparation and cleaning, production line support andMore Product Information