MICROCHEM® 4000, 406 Chemical Resistant Boot Covers
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GR40W-00406-00
ANSEGR40W-00406-00EA 9520 DKK
ANSEGR40W-00406-00
MICROCHEM® 4000, 406 Chemical Resistant Boot Covers
Engangsbeklædning
These boot covers are made from a multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film) that is renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
  • Elastic to top of boot
  • Tie fastening
  • Ultrasonically welded seams
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture.
Certificeringer: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5