Formaldehyd solution 4%

Leverandør: Ampliqon A/S
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

92555.1000-001 92555.1000-003 92555.1000-010
AMPQ92555.1000-001EA 3300 DKK
AMPQ92555.1000-001 AMPQ92555.1000-003 AMPQ92555.1000-010
Formaldehyd solution 4%
Formaldehyd

(10% Formaline solution)

Formel: CH₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR