Universal reference RNAs
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
740000 740100 740200
AGLS740000EA 7107 DKK
AGLS740000 AGLS740100 AGLS740200
Universal reference RNAs
Nukleinsyrereagenser RNAi
High-quality total RNA for human, mouse, and rat microarray gene-expression profiling that acts as a consistent control for standard data set comparisons.
  • Comprised of 10 cell lines for human, 11 for mouse and 14 for rat giving broad gene coverage
  • UHRR selected as one of the two standards in the FDA led Microarray Quality Control (MAQC) project
The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.
Leveringsinformation: Includes RNase-free water, 1,5 ml.
Advarsel: For research use only. Not for use in diagnostic procedures.
Bestil nu
Specifikationer
Environmentally Preferable
Ansøgning