Restriction enzymes, Dpn I restriction enzymes
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
500402
AGLS500402EA 1187 DKK
AGLS500402
Restriction enzymes, Dpn I restriction enzymes
Nukleinsyrereagenser Restriction Enzymes
Dpn I is an ideal enzyme for cloning and southern blot analysis.
  • High purity enzyme preparation
Leveringsinformation: Includes 10X universal reaction buffer.
Advarsel: For research use only. Not for use in diagnostic procedures.