Benchtop cooler, StrataCooler LP

Leverandør: AGILENT
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

401349
AGLS401349EA 1701 DKK
AGLS401349
Benchtop cooler, StrataCooler LP
Kølere Bordkøler
The StrataCooler LP benchtop cooler provides the most protection for enzymes whether in freezer or on benchtop.

  • Maintains –15 °C for at least 2 hours
  • Eliminates ice chips and potential contamination

Cooler with slots for holding 1,5 ml and 0,5 ml tubes to keep temperature-sensitive enzymes cold while working at the bench.

Leveringsinformation: Cooler includes chilled lid, adapters and a locking handle.
Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR