Dilator set, Aesculap®
Leverandør: Aesculap
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EM099R EM100R EM101R EM102R EM103R EM104R EM105R EM106R EM107R EM108R EM109R EM110R EM111R EM112R EM113R EM114R EM115R EM116R EM117R EM118R EM119R EM120R EM121R EM122R EM123R EM124R EM125R EM126R EM194R JG170R
AESCEM099REA 0 DKK
AESCEM099R AESCEM100R AESCEM101R AESCEM102R AESCEM103R AESCEM104R AESCEM105R AESCEM106R AESCEM107R AESCEM108R AESCEM109R AESCEM110R AESCEM111R AESCEM112R AESCEM113R AESCEM114R AESCEM115R AESCEM116R AESCEM117R AESCEM118R AESCEM119R AESCEM120R AESCEM121R AESCEM122R AESCEM123R AESCEM124R AESCEM125R AESCEM126R AESCEM194R AESCJG170R
Dilator set, Aesculap®
Dilatorer
Complete set of uterine dilators.
Leveringsinformation: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).