Membrane filters, mixed cellulose ester, black discs
Leverandør: Advantech
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A045N047A A045R047W
ADVTA045N047AEA 2530 DKK
ADVTA045N047A ADVTA045R047W
Membrane filters, mixed cellulose ester, black discs
Filtre Membranfiltre
Standard membranes for many laboratory applications including filter sterilising biological fluids, microbiology, contamination analysis and air monitoring.
  • High porosity provides superior flow rate
  • High purity and triton-free
  • Rapid wetting time
  • Non reactive to pyrogens
These membranes are composed of cellulose acetate and cellulose nitrate. These filters withstand autoclaving temperatures up to 130 °C without adversely affecting bubble point, flow rate or microbiological recovery.