Ammonia - Ammonium Chloride Buffer Solution, pH 10
Leverandør: CPACHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CH8004008.1L
ACSDCH8004008.1LEA 697 DKK
ACSDCH8004008.1L
Ammonia - Ammonium Chloride Buffer Solution, pH 10
Buffere pH buffere
Ammonia - Ammonium chloride buffer solution.
  • Shelf life: 12 months
  • Storage temperature: 15 to 25 °C
Certificeringer: ISO 9001