Svovlsyre diluted

Leverandør: CPACHEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

CH8002119.1L
ACSDCH8002119.1LEA 549 DKK
ACSDCH8002119.1L
Svovlsyre diluted
Svovlsyre

Certificeringer: ISO 9001

Formel: H₂SO₄
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00064589
CAS-nummer: 7664-93-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR