Potassium pyroantimonate, Test Solution (TS)

Leverandør: CPACHEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassium hexahydroxoantimonate, Potassium hexahydroxoantimonate(V), Potassium antimonate

CH8002106.L1
ACSDCH8002106.L1EA 514 DKK
ACSDCH8002106.L1
Potassium pyroantimonate, Test Solution (TS)
Potassium pyroantimonate

The standard is specially intended for use in testing ChP and monographs. This solution was produced according to the specifications using ChP reagent quality components and water meeting the ChP monograph requirement for Purified Water.


Bestillingsoplysninger: Shelf Life on Ship Date: 12 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.


Advarsel: The solution bottle should be open for the minimum time required. After use, the bottle should be tightly recapped and stored under normal laboratory conditions.

Formel: KSbO₃·3H₂O
MDL Number: MFCD00011415
CAS-nummer: 12208-13-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR