Tungsten Standard Solution, 1000 mg/L W in Water (from W), Standard for ICP
Leverandør: CPACHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C066.W.L1
ACSDC066.W.L1EA 783 DKK
ACSDC066.W.L1
Tungsten Standard Solution, 1000 mg/L W in Water (from W), Standard for ICP
Standarder Kationiske standarder Tungsten Standards
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certificeringer: ISO 17034, 17025, 9001 certified.