Kaliumhydrogensulfat 99%, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potassium bisulphate

447610025 447610100 447615000
ACRO447610025EA 1700 DKK
ACRO447610025 ACRO447610100 ACRO447615000
Kaliumhydrogensulfat 99%, extra pure
Potassium hydrogen sulphate
Formel: KHSO₄
Smeltepunkt: 214 °C
Massefylde: 2,32 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011404
CAS-nummer: 7646-93-7
UN nummer: 2509
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Solubility (10 % in water) Clear colorless
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.002 %
Ammonium (NH4) ≤0.003 %
Iron (Fe) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR