2-Hydroxy-6-methoxybenzoesyre 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid

437670100
ACRO437670100EA 1130 DKK
ACRO437670100
2-Hydroxy-6-methoxybenzoesyre 98%
2-Hydroxy-6-methoxybenzoesyre
Formel: C₈H₈O₄
Molekylvægt: 168.15 g/mol
Smeltepunkt: 135.9…136.4 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00674090
CAS-nummer: 3147-64-6

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 134 to 138 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR