Natriumdiethyldithiocarbamat trihydrat ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

424311000 424315000
ACRO424311000EA 540 DKK
ACRO424311000 ACRO424315000
Natriumdiethyldithiocarbamat trihydrat ACS
Natriumdiethyldithiocarbamat trihydrat
Formel: (C₂H₅)₂NCSSNa·3H₂O
Massefylde: 1,1 g/cm³ (20 °C)
MDL Number: MFCD00150617
CAS-nummer: 20624-25-3
EINECS: 205-710-6
UN nummer: 3077
ADR: 9,III
Merck Index: 12,03443

Bestil nu

Specifikationer Testresultater

Appearance Off-white to light grey Powder or crystals
Infrared spectrum Conforms
Solubility (in water) Passes test
Sensitivity (to Copper) Passes test
Sodium (Na) 30.5 to 32.5 % (as Na2SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR