4-Methyl-2-pentanon 98.5+% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methylisobutylketon, Isobutylmethylketon

423960010 423965000 423960025
ACRO423960010EA 600 DKK
ACRO423960010 ACRO423965000 ACRO423960025
4-Methyl-2-pentanon 98.5+% ACS
4-Methyl-2-pentanone
Formel: (CH₃)₂CHCH₂COCH₃
Kogepunkt: 117,4 °C (1013 hPa)
Smeltepunkt: –84 °C
Massefylde: 0,8 g/cm³
Flammepunkt: 14 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008938
CAS-nummer: 108-10-1
EINECS: 203-550-1
UN nummer: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3970 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %
Titrable acid ≤0.002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR