Ammoniumnitrat 98% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

423350010 423350250 423350050
ACRO423350010EA 549 DKK
ACRO423350010 ACRO423350250 ACRO423350050
Ammoniumnitrat 98% ACS
Ammonium nitrate
Formel: NH₄NO₃
Kogepunkt: 230 °C (1013 hPa)
Smeltepunkt: 169 °C
Massefylde: 1,72 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011425
CAS-nummer: 6484-52-2
EINECS: 229-347-8
UN nummer: 1942
ADR: 5.1,III
Merck Index: 13,00538

Bestil nu

Specifikationer Testresultater

Appearance White Adhering crystals
Residue after ignition ≤0.01 %
pH 4.5 to 6.0 (5 % at 25°C)
Titration excess NaOH ≥97.5 %
Chloride (Cl) ≤5 ppm
Nitrite (NO2) (Limit about 5 ppm) Passes test
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR