pH Indicator solution universal in isopropanol (IPA), pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

422435000
ACRO422435000EA 894 DKK
ACRO422435000
pH Indicator solution universal in isopropanol (IPA), pure
pH Indicator solution
Flammepunkt: 19 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00148741
UN nummer: 1993
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Solution
Visual transition interval pH 9.0 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR