Natriumthiocyanat 98+% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Sodium rhodanide

419670050 419675000 419670025 419671000
ACRO419670050EA 206 DKK
ACRO419670050 ACRO419675000 ACRO419670025 ACRO419671000
Natriumthiocyanat 98+% ACS
Sodium thiocyanate
Formel: NaSCN
Smeltepunkt: 287 °C
Massefylde: 1,735 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011123
CAS-nummer: 540-72-7
EINECS: 208-754-4
Merck Index: 13,09401

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder or crystals and/or chunks
Titration Argentometric ≥98.0 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Ammonium (NH4) ≤20 ppm
Iron (Fe) ≤2 ppm
Sulfur (S) ≤10 ppm
Heavy metals (as Pb) ≤5 ppm
Impurity ≤0.2 % Carbonate (as Na2CO3)
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR