Paraffin wax, granular, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Wax Paraffin, Paraffin wax

416770020 416770100
ACRO416770020EA 802 DKK
ACRO416770020 ACRO416770100
Paraffin wax, granular, pure
Paraffin
Kogepunkt: 322 °C (760 mmHg)
Smeltepunkt: 58…62 °C
Flammepunkt: 198 °C
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 8002-74-2

Bestil nu

Specifikationer Testresultater

Appearance White Pellets
Infrared spectrum Conforms
Congealing point 58 °C to 62 °C
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR