Cleland's reagens 98%, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

409190010 409190050 409190250 409191000
ACRO409190010EA 395 DKK
ACRO409190010 ACRO409190050 ACRO409190250 ACRO409191000
Cleland's reagens 98%, pure
Cleland's reagens
Formel: C₄H₁₀O₂S₂
Kogepunkt: 125…130 °C (2 mmHg)
Smeltepunkt: 39…44 °C
Flammepunkt: >110 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00004877
CAS-nummer: 3483-12-3
EINECS: 222-468-7
UN nummer: 3335
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Infrared spectrum Conforms
Melting point 38 to 43 °C
Titration Iodimetric 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR