Bromphenol blå natriumsalt ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

403160100 403160050 403160500
ACRO403160100EA 452 DKK
ACRO403160100 ACRO403160050 ACRO403160500
Bromphenol blå natriumsalt ACS
Bromphenol blå natriumsalt
Formel: C₁₉H₉Br₄NaO₅S
Smeltepunkt: >300 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013793
CAS-nummer: 34725-61-6
EINECS: 252-170-2

Bestil nu

Specifikationer Testresultater

Appearance Dark green to brown-black to blue-black Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.0 (yellow) to pH 4.6 (blue)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR