(R)-(+)-1,2-Epoxypropan 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R)-(+)-Propylenoxid, (R)-(+)-Methyloxiran

399570050 399570250
ACRO399570050EA 692 DKK
ACRO399570050 ACRO399570250
(R)-(+)-1,2-Epoxypropan 98+%
(R)-(+)-1,2-Epoxypropan
Formel: C₃H₆O
Smeltepunkt: –112 °C
Massefylde: 0,829 g/cm³
Flammepunkt: –37 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00066211
CAS-nummer: 15448-47-2
UN nummer: 1280
ADR: 3,I

Bestil nu

Specifikationer Testresultater

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3680 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation +12° to +15° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR