3-Iodbenzaldehyd 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

392070010 392070050
ACRO392070010EA 718 DKK
ACRO392070010 ACRO392070050
3-Iodbenzaldehyd 99%
3-Iodobenzaldehyde
Formel: C₇H₅IO
Molekylvægt: 232.02 g/mol
Smeltepunkt: 57…60 °C
MDL Number: MFCD00039573
CAS-nummer: 696-41-3

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥98.5 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR