Glycin 99% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH

385902500 385900500
ACRO385902500EA 509 DKK
ACRO385902500 ACRO385900500
Glycin 99% ACS
Glycin
Formel: NH₂CH₂COOH
Kogepunkt: 181 °C (1013 hPa)
Smeltepunkt: 240 °C
Massefylde: 1,595 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008131
CAS-nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.1 %
Titration with HClO4 ≥98.5 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.005 %
Heavy metals (as Pb) ≤0.002 %
Hydrolyzable substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR