Xylen 98%, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethylbenzene

383930050 383930010 383930250
ACRO383930050EA 927 DKK
ACRO383930050 ACRO383930010 ACRO383930250
Xylen 98%, pure
Xylen
Formel: C₆H₄(CH₃)₂
Kogepunkt: 136…140 °C (760 mmHg)
Smeltepunkt: –34 °C
Massefylde: 0,865 g/cm³
Flammepunkt: 23 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077264
CAS-nummer: 1330-20-7
EINECS: 215-535-7
UN nummer: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥97 % (isomers and ethylbenzene)
Residue after evaporation ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR