L(+)-Asparagin 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH

371601000 371605000 371600025
ACRO371601000EA 885 DKK
ACRO371601000 ACRO371605000 ACRO371600025
L(+)-Asparagin 99%
L(+)-Asparagin
Formel: H₂NCOCH₂CH(NH₂)CO₂H
Molekylvægt: 132.12 g/mol
Kogepunkt: 343
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064401
CAS-nummer: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Specific optical rotation +33° to +36° (20°C, 589 nm) (c=10, 2N HCl)
Titration with HClO4 ≥98.5 % (On dry substance)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR