Kieselgel, Pore size: 40 Å 40-63 µm, Ultrapure for column chromatography

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

360031000 360035000
ACRO360031000EA 446 DKK
ACRO360031000 ACRO360035000
Kieselgel, Pore size: 40 Å 40-63 µm, Ultrapure for column chromatography
Silica gel
Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 7631-86-9

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Particle size 40 to 63 µm
Specific surface area ≥450 m²/g (BET)
Pore volume 0.55 to 0.75 ml/g
pH 5.0 to 8.0 (5 % w/w suspension)
Volatile matter ≤4.0 % (160°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR