Kieselgel 3-6 mm, non toxic for drying

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

357400010 357400050 357400250
ACRO357400010EA 537 DKK
ACRO357400010 ACRO357400050 ACRO357400250
Kieselgel 3-6 mm, non toxic for drying
Silica gel
Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 7631-86-9
REACH: 01-2119379499-16

Bestil nu

Specifikationer Testresultater

Appearance Colorless to white Irregular granules
Particle size 3.0 to 6.0 mm
Residual water ≤2.0 % (140°C, 4 h)
Water adsorption ≥20.0 % (40% rH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR