Dichlormethan 99.9% stabilised for peptide synthesis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methylenchlorid

Stabilization: stabiliseret med amylen

354800025 354800100
ACRO354800025EA 1340 DKK
ACRO354800025 ACRO354800100
Dichlormethan 99.9% stabilised for peptide synthesis
Dichloromethane
Formel: CH₂Cl₂
Kogepunkt: 39…40 °C (760 mmHg)
Smeltepunkt: –97 °C
Massefylde: 1,325 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
UN nummer: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Water ≤0.005 %
GC ≥99.9 %
Color scale ≤10 APHA
Stabilizer 0.003 to 0.006 % (Amylene)
Residue after evaporation ≤0.0005 %
Free amine ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR